2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene
Supplier: Matrix Scientific
Synonyms:
2,4-Bis(chloromethyl)mesitylene
063558-500MG
101925-954EA
146.15
USD
101925-954
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene
2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene
Hazard: Irritant
Formula:
C₁₁H₁₄Cl₂ |
MDL Number:
MFCD00013681 CAS Number: 1585-17-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...