3-{[2-(Trifluoromethyl)phenyl]amino}cyclohex-2-en-1-one

Supplier: Matrix Scientific

063557-500MG
101925-952EA 484.62 USD
101925-952
3-{[2-(Trifluoromethyl)phenyl]amino}cyclohex-2-en-1-one
3-{[2-(Trifluoromethyl)phenyl]amino}cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₃H₁₂F₃NO
MDL Number: MFCD15146485
CAS Number: 1217862-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR