2,4,6-Triethyl-3-(hydroxymethyl)benzaldehyde oxime

Supplier: Matrix Scientific

063554-500MG
101925-946EA 484.62 USD
101925-946
2,4,6-Triethyl-3-(hydroxymethyl)benzaldehyde oxime
2,4,6-Triethyl-3-(hydroxymethyl)benzaldehyde oxime

Hazard: Irritant

Formula: C₁₄H₂₁NO₂
MDL Number: MFCD00757575
CAS Number: 347367-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR