2,3,5,6-Tetramethylterephthalaldehyde

Supplier: Matrix Scientific

063553-500MG
101925-944EA 443.08 USD
101925-944
2,3,5,6-Tetramethylterephthalaldehyde
2,3,5,6-Tetramethylterephthalaldehyde

Hazard: Irritant

Formula: C₁₂H₁₄O₂
Melting Pt: 165 °C
MDL Number: MFCD00194529
CAS Number: 7072-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR