3-[(2-Methylphenyl)amino]cyclohex-2-en-1-one
Supplier: Matrix Scientific
063552-500MG
101925-942EA
484.62
USD
101925-942
3-[(2-Methylphenyl)amino]cyclohex-2-en-1-one
3-[(2-Methylphenyl)amino]cyclohex-2-en-1-one
Hazard: Irritant
Formula:
C₁₃H₁₅NO |
MDL Number:
MFCD15146483 CAS Number: 400873-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...