3-[(2-Methyl-5-nitrophenyl)amino]cyclohex-2-en-1-one

Supplier: Matrix Scientific

063547-500MG
101925-932EA 484.62 USD
101925-932
3-[(2-Methyl-5-nitrophenyl)amino]cyclohex-2-en-1-one
3-[(2-Methyl-5-nitrophenyl)amino]cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₃
MDL Number: MFCD15146480
CAS Number: 1217862-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR