3-[(4-Chlorophenyl)amino]cyclohex-2-en-1-one

Supplier: Matrix Scientific

063543-500MG
101925-924EA 484.62 USD
101925-924
3-[(4-Chlorophenyl)amino]cyclohex-2-en-1-one
3-[(4-Chlorophenyl)amino]cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₂H₁₂ClNO
MDL Number: MFCD00129271
CAS Number: 36646-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR