3-[(3-Chloro-4-methylphenyl)amino]cyclohex-2-en-1-one

Supplier: Matrix Scientific

063540-500MG
101925-918EA 484.62 USD
101925-918
3-[(3-Chloro-4-methylphenyl)amino]cyclohex-2-en-1-one
3-[(3-Chloro-4-methylphenyl)amino]cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₃H₁₄ClNO
MW: 235.72 g/mol
MDL Number: MFCD15146477
CAS Number: 1217862-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR