3-[(4-Methoxyphenyl)amino]-5,5-dimethylcyclohex-2-en-1-one

Supplier: Matrix Scientific

063538-500MG
101925-914EA 484.62 USD
101925-914
3-[(4-Methoxyphenyl)amino]-5,5-dimethylcyclohex-2-en-1-one
3-[(4-Methoxyphenyl)amino]-5,5-dimethylcyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₅H₁₉NO₂
MDL Number: MFCD00128469
CAS Number: 24706-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR