3-[(4-Fluorophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one

Supplier: Matrix Scientific

063537-500MG
101925-912EA 484.62 USD
101925-912
3-[(4-Fluorophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one
3-[(4-Fluorophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₄H₁₆FNO
MDL Number: MFCD00128471
CAS Number: 100445-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR