3-[(5-Methylisoxazol-3-yl)amino]cyclohex-2-en-1-one

Supplier: Matrix Scientific

063529-500MG
101925-896EA 606.15 USD
101925-896
3-[(5-Methylisoxazol-3-yl)amino]cyclohex-2-en-1-one
3-[(5-Methylisoxazol-3-yl)amino]cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
MDL Number: MFCD15146473
CAS Number: 1217863-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR