3-[(2,3-Dichlorophenyl)amino]cyclohex-2-en-1-one

Supplier: Matrix Scientific

063525-500MG
101925-888EA 484.62 USD
101925-888
3-[(2,3-Dichlorophenyl)amino]cyclohex-2-en-1-one
3-[(2,3-Dichlorophenyl)amino]cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₂H₁₁Cl₂NO
MDL Number: MFCD01735081
CAS Number: 145657-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR