3-(1-Naphthylamino)cyclohex-2-en-1-one

Supplier: Matrix Scientific

063523-500MG
101925-884EA 484.62 USD
101925-884
3-(1-Naphthylamino)cyclohex-2-en-1-one
3-(1-Naphthylamino)cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₆H₁₅NO
MDL Number: MFCD01168839
CAS Number: 326610-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR