3-[(4-Chlorophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one

Supplier: Matrix Scientific

063517-500MG
101925-872EA 484.62 USD
101925-872
3-[(4-Chlorophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one
3-[(4-Chlorophenyl)amino]-5,5-dimethylcyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₄H₁₆ClNO
MDL Number: MFCD00128477
CAS Number: 24706-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR