3-[(2-Fluorophenyl)amino]cyclohex-2-en-1-one

Supplier: Matrix Scientific

063514-500MG
101925-866EA 484.62 USD
101925-866
3-[(2-Fluorophenyl)amino]cyclohex-2-en-1-one
3-[(2-Fluorophenyl)amino]cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₂H₁₂FNO
MDL Number: MFCD00042277
CAS Number: 124907-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR