Thiophene-2-carboxaldoxime

Supplier: Matrix Scientific

063508-500MG
101925-854EA 183.08 USD
101925-854
Thiophene-2-carboxaldoxime
Thiophene-2-carboxaldoxime

Hazard: Irritant

Formula: C₅H₅NOS
MW: 127.17 g/mol
MDL Number: MFCD00022509
CAS Number: 29683-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR