Thiophene-3-carboxaldoxime
Supplier: Matrix Scientific
063507-500MG
101925-852EA
193.85
USD
101925-852
Thiophene-3-carboxaldoxime
Thiophene-3-carboxaldoxime
Hazard: Irritant
Formula:
C₅H₅NOS |
MDL Number:
MFCD00151853 CAS Number: 42466-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...