Ethyl-3-(2-oxoazepan-1-yl)propanoate

Supplier: Matrix Scientific

063504-500MG
101925-848EA 364.62 USD
101925-848
Ethyl-3-(2-oxoazepan-1-yl)propanoate
Ethyl-3-(2-oxoazepan-1-yl)propanoate

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MDL Number: MFCD11652348
CAS Number: 88948-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR