2-[(4-Nitro-1H-pyrazol-3-yl)thio]-1,3-benzothiazole

Supplier: Matrix Scientific

063498-500MG
101925-836EA 364.62 USD
101925-836
2-[(4-Nitro-1H-pyrazol-3-yl)thio]-1,3-benzothiazole
2-[(4-Nitro-1H-pyrazol-3-yl)thio]-1,3-benzothiazole

Hazard: Irritant

Formula: C₁₀H₆N₄O₂S₂
MDL Number: MFCD15146460
CAS Number: 1217862-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR