4-(3-Methyl-4-nitro-1H-pyrazol-5-yl)morpholine

Supplier: Matrix Scientific

063497-500MG
101925-834EA 484.62 USD
101925-834
4-(3-Methyl-4-nitro-1H-pyrazol-5-yl)morpholine
4-(3-Methyl-4-nitro-1H-pyrazol-5-yl)morpholine

Hazard: Irritant

Formula: C₈H₁₂N₄O₃
MDL Number: MFCD00459267
CAS Number: 53960-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR