[1H-Indol-3-yl(phenyl)methyl]methylamine

Supplier: Matrix Scientific

063495-500MG
101925-830EA 484.62 USD
101925-830
[1H-Indol-3-yl(phenyl)methyl]methylamine
[1H-Indol-3-yl(phenyl)methyl]methylamine

Hazard: Irritant

Formula: C₁₆H₁₆N₂
MDL Number: MFCD03004606
CAS Number: 33268-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR