2-Chloro-4-methylquinoline-3-carbonitrile
Supplier: Matrix Scientific
063478-500MG
101925-798EA
446.15
USD
101925-798
2-Chloro-4-methylquinoline-3-carbonitrile
2-Chloro-4-methylquinoline-3-carbonitrile
Hazard: Irritant
Formula:
C₁₁H₇ClN₂ |
MDL Number:
MFCD00833327 CAS Number: 101617-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...