4-Methyl-2-oxo-1,2-dihydroquinoline-3-carbonitrile

Supplier: Matrix Scientific

063477-500MG
101925-796EA 336.92 USD
101925-796
4-Methyl-2-oxo-1,2-dihydroquinoline-3-carbonitrile
4-Methyl-2-oxo-1,2-dihydroquinoline-3-carbonitrile

Hazard: Irritant

Formula: C₁₁H₈N₂O
Melting Pt: >300 °C
MDL Number: MFCD00833317
CAS Number: 28448-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR