Acetoxyacetyl chloride

Supplier: Matrix Scientific

Synonyms: Acetoxyacetylchloride2-Chloro-2-oxoethyl acetate

063473-100G 063473-25G
101925-786EA 60 USD
101925-786 101925-788
Acetoxyacetyl chloride
Acetoxyacetyl chloride

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₄H₅ClO₃
MW: 136.53 g/mol
Boiling Pt: 55 °C (12 mmHg)
Density: 1.27
Flash Pt: 71 °C
MDL Number: MFCD00011535
CAS Number: 13831-31-7
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR