4-Phenyl-1,3-thiazole-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 4-Phenylthiazole-2-carboxylic acid

063472-500MG
101925-784EA 498.46 USD
101925-784
4-Phenyl-1,3-thiazole-2-carboxylic acid
4-Phenyl-1,3-thiazole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₂S
MW: 205.24 g/mol
MDL Number: MFCD00990415
CAS Number: 59020-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR