[2-(2-Pyridin-4-yl-1,3-thiazol-4-yl)ethyl]amine

Supplier: Matrix Scientific

063470-500MG
101925-780EA 364.62 USD
101925-780
[2-(2-Pyridin-4-yl-1,3-thiazol-4-yl)ethyl]amine
[2-(2-Pyridin-4-yl-1,3-thiazol-4-yl)ethyl]amine

Hazard: Irritant

Formula: C₁₀H₁₁N₃S
MDL Number: MFCD09724097
CAS Number: 938283-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR