3-Isopropyl-1,2,4-oxadiazol-5-amine

Supplier: Matrix Scientific

063468-500MG
101925-776EA 438.46 USD
101925-776
3-Isopropyl-1,2,4-oxadiazol-5-amine
3-Isopropyl-1,2,4-oxadiazol-5-amine

Hazard: Irritant

Formula: C₅H₉N₃O
MDL Number: MFCD11847816
CAS Number: 3874-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR