3-Bromo-5-(ethylthio)-1H-1,2,4-triazole

Supplier: Matrix Scientific

063464-500MG
101925-768EA 484.62 USD
101925-768
3-Bromo-5-(ethylthio)-1H-1,2,4-triazole
3-Bromo-5-(ethylthio)-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₄H₆BrN₃S
MDL Number: MFCD15146451
CAS Number: 1209935-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR