3-Cyclopentyl-1H-1,2,4-triazole

Supplier: Matrix Scientific

063459-500MG
101925-758EA 484.62 USD
101925-758
3-Cyclopentyl-1H-1,2,4-triazole
3-Cyclopentyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₇H₁₁N₃
MDL Number: MFCD13188605
CAS Number: 1210317-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR