[(5-Isopropyl-1,2,4-oxadiazol-3-yl)methyl]amine

Supplier: Matrix Scientific

063451-500MG
101925-742EA 484.62 USD
101925-742
[(5-Isopropyl-1,2,4-oxadiazol-3-yl)methyl]amine
[(5-Isopropyl-1,2,4-oxadiazol-3-yl)methyl]amine

Hazard: Irritant

Formula: C₆H₁₁N₃O
MDL Number: MFCD09971174
CAS Number: 936940-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR