(4,6-Dimethyl-3-nitro-2-oxopyridin-1(2H)-yl)acetic acid

Supplier: Matrix Scientific

063445-500MG
101925-730EA 484.62 USD
101925-730
(4,6-Dimethyl-3-nitro-2-oxopyridin-1(2H)-yl)acetic acid
(4,6-Dimethyl-3-nitro-2-oxopyridin-1(2H)-yl)acetic acid

Hazard: Irritant

Formula: C₉H₁₀N₂O₅
MDL Number: MFCD02752891
CAS Number: 367454-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR