1-(4-Methylphenyl)pyrrolidin-2-one

Supplier: Matrix Scientific

063441-500MG
101925-722EA 484.62 USD
101925-722
1-(4-Methylphenyl)pyrrolidin-2-one
1-(4-Methylphenyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₁₁H₁₃NO
MDL Number: MFCD00138487
CAS Number: 3063-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR