Methyl-3-(2-oxopyrrolidin-1-yl)benzoate

Supplier: Matrix Scientific

063438-500MG
101925-716EA 484.62 USD
101925-716
Methyl-3-(2-oxopyrrolidin-1-yl)benzoate
Methyl-3-(2-oxopyrrolidin-1-yl)benzoate

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MDL Number: MFCD00593013
CAS Number: 329205-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR