1-(3-Methylphenyl)pyrrolidin-2-one

Supplier: Matrix Scientific

063437-500MG
101925-714EA 484.62 USD
101925-714
1-(3-Methylphenyl)pyrrolidin-2-one
1-(3-Methylphenyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₁₁H₁₃NO
MDL Number: MFCD00746351
CAS Number: 24059-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR