5-(3-Methoxyphenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione

Supplier: Matrix Scientific

063434-500MG
101925-708EA 183.08 USD
101925-708
5-(3-Methoxyphenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione
5-(3-Methoxyphenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione

Hazard: Irritant

Formula: C₉H₉N₃OS
MDL Number: MFCD01122586
CAS Number: 419540-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR