5-(4-Nitrophenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione

Supplier: Matrix Scientific

063432-500MG
101925-704EA 484.62 USD
101925-704
5-(4-Nitrophenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione
5-(4-Nitrophenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione

Hazard: Irritant

Formula: C₈H₆N₄O₂S
MDL Number: MFCD00553303
CAS Number: 6898-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR