1-Allyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde

Supplier: Matrix Scientific

063431-500MG
101925-702EA 243.08 USD
101925-702
1-Allyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
1-Allyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₉H₁₂N₂O
MDL Number: MFCD12103568
CAS Number: 1155595-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR