8,9,10,11-Tetrahydro[1]benzothieno[3,2-e]tetrazolo[1,5-c]pyrimidine

Supplier: Matrix Scientific

063428-500MG
101925-696EA 606.15 USD
101925-696
8,9,10,11-Tetrahydro[1]benzothieno[3,2-e]tetrazolo[1,5-c]pyrimidine
8,9,10,11-Tetrahydro[1]benzothieno[3,2-e]tetrazolo[1,5-c]pyrimidine

Hazard: Irritant

Formula: C₁₀H₉N₅S
MW: 231.28 g/mol
MDL Number: MFCD00454418
CAS Number: 40277-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR