6-Nitro-2-(2-phenylethyl)-1H-benzimidazole

Supplier: Matrix Scientific

063422-500MG
101925-682EA 484.62 USD
101925-682
6-Nitro-2-(2-phenylethyl)-1H-benzimidazole
6-Nitro-2-(2-phenylethyl)-1H-benzimidazole

Hazard: Irritant

Formula: C₁₅H₁₃N₃O₂
MDL Number: MFCD00468700
CAS Number: 330592-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR