Methyl-1-methylhydrazinecarboxylate

Supplier: Matrix Scientific

063418-500MG
101925-676EA 253.85 USD
101925-676
Methyl-1-methylhydrazinecarboxylate
Methyl-1-methylhydrazinecarboxylate

Hazard: Irritant

Formula: C₃H₈N₂O₂
MDL Number: MFCD15146446
CAS Number: 20628-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR