Methyl-3-hydroxypropanoate

Synonyms: Methyl 3-hydroxypropanoate

063393-500MG
101925-622EA 253.47 USD
101925-622
Methyl-3-hydroxypropanoate
Methyl-3-hydroxypropanoate

Hazard: Irritant

Formula: C₄H₈O₃
MW: 104.11 g/mol
MDL Number: MFCD00272293
CAS Number: 6149-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR