2-(4-Bromo-1H-pyrazol-1-yl)propanamide

Supplier: Matrix Scientific

063381-500MG
101925-614EA 484.62 USD
101925-614
2-(4-Bromo-1H-pyrazol-1-yl)propanamide
2-(4-Bromo-1H-pyrazol-1-yl)propanamide

Hazard: Irritant

Formula: C₆H₈BrN₃O
MDL Number: MFCD12824359
CAS Number: 1183357-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR