4-Bromo-1-isopropyl-3-methyl-1H-pyrazole

Supplier: Matrix Scientific

063378-500MG
101925-610EA 484.62 USD
101925-610
4-Bromo-1-isopropyl-3-methyl-1H-pyrazole
4-Bromo-1-isopropyl-3-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₇H₁₁BrN₂
MDL Number: MFCD14702926
CAS Number: 1215295-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR