4-Chloro-5-methyl-1H-pyrazole

Supplier: Matrix Scientific

063368-500MG
101925-602EA 484.62 USD
101925-602
4-Chloro-5-methyl-1H-pyrazole
4-Chloro-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₄H₅ClN₂
MW: 116.55 g/mol
MDL Number: MFCD00514185
CAS Number: 15878-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR