2-(2-Methyl-1,3-thiazol-4-yl)acetamide

Supplier: Matrix Scientific

063352-500MG
101925-596EA 364.62 USD
101925-596
2-(2-Methyl-1,3-thiazol-4-yl)acetamide
2-(2-Methyl-1,3-thiazol-4-yl)acetamide

Hazard: Irritant

Formula: C₆H₈N₂OS
MW: 156.21 g/mol
MDL Number: MFCD03011393
CAS Number: 185623-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR