tert-Butyl-4-(ethoxymethyl)piperidine-1-carboxylate

Supplier: Matrix Scientific

063349-500MG
101925-590EA 484.62 USD
101925-590
tert-Butyl-4-(ethoxymethyl)piperidine-1-carboxylate
tert-Butyl-4-(ethoxymethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₅NO₃
MW: 243.35 g/mol
MDL Number: MFCD15143207
CAS Number: 247132-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR