(1Z)-2-(3-Chlorophenyl)-N'-cyanoethanimidamide

Supplier: Matrix Scientific

063341-500MG
101925-574EA 484.62 USD
101925-574
(1Z)-2-(3-Chlorophenyl)-N'-cyanoethanimidamide
(1Z)-2-(3-Chlorophenyl)-N'-cyanoethanimidamide

Hazard: Irritant

Formula: C₉H₈ClN₃
MDL Number: MFCD15146425
CAS Number: 78622-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR