8-Ethyl-2,8-diazaspiro[4.5]decane

Supplier: Matrix Scientific

063328-500MG
101925-554EA 606.15 USD
101925-554
8-Ethyl-2,8-diazaspiro[4.5]decane
8-Ethyl-2,8-diazaspiro[4.5]decane

Hazard: Irritant

Formula: C₁₀H₂₀N₂
MDL Number: MFCD15146422
CAS Number: 1217862-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR