8-Methyl-2-azaspiro[4.5]decane

Supplier: Matrix Scientific

063327-500MG
101925-552EA 606.15 USD
101925-552
8-Methyl-2-azaspiro[4.5]decane
8-Methyl-2-azaspiro[4.5]decane

Hazard: Irritant

Formula: C₁₀H₁₉N
MDL Number: MFCD15146421
CAS Number: 1217862-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR