2-Azaspiro[4.5]decane

Supplier: Matrix Scientific

063326-500MG
101925-550EA 353.85 USD
101925-550
2-Azaspiro[4.5]decane
2-Azaspiro[4.5]decane

Hazard: Irritant

Formula: C₉H₁₇N
MDL Number: MFCD06656114
CAS Number: 176-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR